Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138517.3 | NP_612526.2 | 4 | 629 | ||
GenBank mRNA | BC127475.1 | - | 4 | 609 | 7124652 | |
FQ220462.1 | - | 1 | 176 | |||
FQ220750.1 | - | 4 | 651 | |||
FQ221957.1 | - | 4 | 651 | |||
FQ228927.1 | - | 4 | 651 |
Gene Symbol: Gzmbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728257.4 | XP_002728303.2 | 4 | 697 |