Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138543.1 | NP_612552.1 | 7 | 503 | ||
XM_008772793.1 | XP_008771015.1 | 7 | 517 | |||
XM_008772794.1 | XP_008771016.1 | 7 | 517 | |||
XM_008772795.1 | XP_008771017.1 | 7 | 517 | |||
GenBank mRNA | AF372654.1 | - | 7 | 503 | ||
AY145898.1 | - | 7 | 522 | |||
BC078733.1 | - | 7 | 444 | |||
BC161837.1 | - | 7 | 476 | 9029236 |