Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou3f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138837.1 | NP_620192.1 | 2 | 2964 | ||
GenBank mRNA | AJ001641.1 | - | 2 | 2964 |