Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138889.2 | NP_620244.1 | 3 | 287 | ||
GenBank mRNA | AF494095.1 | - | 3 | 284 | ||
BC085699.1 | - | 3 | 287 | 7106186 |