Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138890.2 | NP_620245.2 | 3, 4 | 901 | ||
XM_006239774.2 | XP_006239836.1 | 4, 5 | 1146 | |||
XM_008764503.1 | XP_008762725.1 | 3, 4 | 747 | |||
GenBank mRNA | AF494093.2 | - | 3, 4 | 901 |