Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318219.1 | NP_001305148.1 | 4 | 494 | ||
NM_007186.5 | NP_009117.2 | 6 | 915 | |||
XM_005260262.4 | XP_005260319.1 | 6 | 668 | |||
XM_005260263.4 | XP_005260320.1 | 6 | 668 | |||
XM_005260264.4 | XP_005260321.1 | 6 | 668 | |||
XM_006723690.3 | XP_006723753.1 | 6 | 665 | |||
XM_006723691.1 | XP_006723754.1 | 6 | 881 | |||
XM_006723692.3 | XP_006723755.1 | 5 | 591 | |||
XM_006723693.3 | XP_006723756.1 | 5 | 575 | |||
XM_006723694.3 | XP_006723757.1 | 6 | 668 | |||
XM_011528517.2 | XP_011526819.1 | 6 | 668 | |||
XM_011528518.2 | XP_011526820.1 | 6 | 667 | |||
XM_017027617.1 | XP_016883106.1 | 6 | 668 | |||
GenBank mRNA | AB621828.1 | - | 6 | 607 | ||
AF022655.1 | - | 4 | 489 | |||
AF049105.1 | - | 7 | 939 | |||
AK297572.1 | - | 5 | 649 | |||
AK307861.1 | - | 6 | 633 | |||
BC071869.1 | - | 6 | 592 | |||
BC144706.1 | - | 4 | 440 |