Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139038.1 | NP_620607.1 | 1 | 262 | ||
XM_008760073.1 | XP_008758295.1 | 544 | ||||
GenBank mRNA | U80076.1 | - | 1 | 262 |
Gene Symbol: LOC100911794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749037.3 | XP_003749085.1 | 1 | 544 | ||
XM_008760195.1 | XP_008758417.1 | 1 | 544 |