Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il11ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139116.1 | NP_620816.1 | 13 | 1557 | ||
XM_006238021.2 | XP_006238083.1 | 12 | 2275 | |||
XM_006238022.2 | XP_006238084.1 | 12 | 1695 | |||
XM_006238023.2 | XP_006238085.1 | 13 | 2177 | |||
GenBank mRNA | AF347936.1 | - | 13 | 1557 | ||
BC070921.1 | - | 13 | 1552 | 7105297 |