Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139325.3 | NP_647541.1 | 12 | 2030 | ||
XM_006237330.2 | XP_006237392.1 | 12 | 2311 | |||
GenBank mRNA | AF019973.1 | - | 12 | 2030 | ||
M11931.1 | - | 12 | 2026 |
Gene Symbol: LOC100911625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750657.3 | XP_003750705.1 | 12 | 2081 | ||
XM_006244636.2 | XP_006244698.1 | 12 | 2313 | |||
GenBank mRNA | FQ212469.1 | - | 1 | 388 |