Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007028.4 | NP_008959.3 | 2 | 403 | ||
NR_134870.1 | 2 | 403 | ||||
NR_134871.1 | 2 | 403 | ||||
XM_011514264.1 | XP_011512566.1 | 2 | 387 | |||
XM_011514265.1 | XP_011512567.1 | 2 | 387 | |||
XM_011514266.1 | XP_011512568.1 | 2 | 387 | |||
GenBank mRNA | AB303153.1 | - | 1 | 277 | ||
AF230386.1 | - | 1 | 386 | |||
AF230387.1 | - | 1 | 386 | |||
AK222729.1 | - | 2 | 387 | |||
AK290270.1 | - | 2 | 390 | |||
BC017017.1 | - | 1 | 364 | 3846513 | ||
BG403536.1 | - | 2 | 400 | |||
BM847963.1 | - | 2 | 385 | |||
BT006675.1 | - | 1 | 277 | |||
CB147821.1 | - | 2 | 387 | |||
DB244000.1 | - | 2 | 387 | |||
Y07828.1 | - | 1 | 386 |