Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153303.2 | NP_695215.2 | 1 | 150 | ||
GenBank mRNA | AY054303.1 | - | 1 | 132 | ||
BC078714.1 | - | 1 | 150 | 7134604 |