Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172021.2 | NP_742018.2 | 3 | 384 | ||
XM_006247266.2 | XP_006247328.1 | 2 | 1534 | |||
XM_008767953.1 | XP_008766175.1 | 2 | 1534 | |||
GenBank mRNA | AJ278800.1 | - | 2 | 370 | ||
BC076503.1 | - | 3 | 384 | 7100584 |