Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172068.1 | NP_742065.1 | 9 | 936 | ||
XM_006233864.1 | XP_006233926.1 | 9 | 947 | |||
XM_006233865.1 | XP_006233927.1 | 8 | 916 | |||
GenBank mRNA | AF182952.1 | - | 9 | 936 |
Gene Symbol: LOC100912008 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233726.2 | XP_006233788.1 | 9 | 947 | ||
XM_006233727.2 | XP_006233789.1 | 916 | ||||
XM_006233728.2 | XP_006233790.1 | 938 |