Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172074.3 | NP_742071.1 | 2 | 876 | ||
XM_006234473.2 | XP_006234535.1 | 3 | 488 | |||
XM_006234474.2 | XP_006234536.1 | 4 | 600 | |||
GenBank mRNA | AF150091.1 | - | 2 | 311 | ||
BC091116.1 | - | 2 | 877 | 7366212 |