Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173102.2 | NP_775125.1 | 4 | 1891 | ||
GenBank mRNA | AB011679.1 | - | 4 | 1850 |