Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gimap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173153.2 | NP_775176.2 | 3 | 275 | ||
GenBank mRNA | AM285343.1 | - | 3 | 275 | ||
AM285683.1 | - | 3 | 275 | |||
AY070268.1 | - | 2 | 257 | |||
BC070952.1 | - | 3 | 263 | 7100695 | ||
CK479589.1 | - | 3 | 275 | |||
DQ125339.1 | - | 3 | 264 | |||
DQ125340.1 | - | 3 | 264 |