Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173308.2 | NP_775430.2 | 3 | 1455 | ||
GenBank mRNA | AJ535753.1 | - | 3 | 1480 | ||
BC079316.1 | - | 3 | 1455 | 7135022 |