Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128203.1 | NP_001121675.1 | 4 | 333 | ||
NM_007069.3 | NP_009000.2 | 3 | 617 | |||
XM_006718426.1 | XP_006718489.1 | 4 | 535 | |||
XM_011544741.1 | XP_011543043.1 | 3 | 519 | |||
GenBank mRNA | AB030814.1 | - | 3 | 243 | ||
AB439591.1 | - | 2 | 194 | |||
AF317086.1 | - | 3 | 248 | |||
AK313075.1 | - | 3 | 616 | |||
AX775893.1 | - | 3 | 248 | |||
BC001387.2 | - | 3 | 549 | 3051149 | ||
BC103807.2 | - | 3 | 338 | 40000132 | ||
BC103808.2 | - | 3 | 288 | |||
BG719903.1 | - | 4 | 328 | |||
CK003948.1 | - | 4 | 334 | |||
X92814.1 | - | 3 | 601 |