Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173840.1 | NP_776212.1 | 11 | 575 | ||
XM_006249160.2 | XP_006249222.1 | 12 | 746 | |||
XM_008769138.1 | XP_008767360.1 | 11 | 1028 | |||
XM_008769139.1 | XP_008767361.1 | 10 | 992 | |||
GenBank mRNA | AY170849.1 | - | 11 | 575 | ||
FQ231408.1 | - | 9 | 435 | |||
FQ235223.1 | - | 9 | 435 |