Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175758.3 | NP_786934.1 | 3 | 1037 | ||
GenBank mRNA | AJ132846.1 | - | 3 | 768 | ||
BC080242.1 | - | 3 | 1037 | 7100054 |