Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcpip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176857.3 | NP_789827.3 | 1 | 528 | ||
GenBank mRNA | AB045378.2 | - | 1 | 528 | ||
AF289091.1 | - | 586 |