Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177931.2 | NP_808792.1 | 3 | 304 | ||
XM_006238515.2 | XP_006238577.1 | 4 | 506 | |||
XM_006238516.2 | XP_006238578.1 | 5 | 619 | |||
XM_006238518.2 | XP_006238580.1 | 4 | 506 | |||
GenBank mRNA | AB105378.1 | - | 3 | 304 |