Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181367.2 | NP_852032.1 | 7 | 1288 | ||
XM_006249914.2 | XP_006249976.1 | 5 | 1780 | |||
XM_006249915.2 | XP_006249977.1 | 5 | 1782 | |||
XM_006249916.2 | XP_006249978.1 | 8 | 1850 | |||
XM_006249917.1 | XP_006249979.1 | 5 | 1141 | |||
GenBank mRNA | AY273890.1 | - | 7 | 1288 | ||
AY724502.1 | - | 7 | 1288 |