Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181372.2 | NP_852037.1 | 1 | 144 | ||
XM_006237081.2 | XP_006237143.1 | 3 | 449 | |||
XM_006237082.2 | XP_006237144.1 | 2 | 531 | |||
XM_006237083.2 | XP_006237145.1 | 1 | 25 | |||
XM_008763311.1 | XP_008761533.1 | 2 | 528 | |||
GenBank mRNA | AF486185.1 | - | 1 | 85 | ||
AF486186.1 | - | 1 | 144 | |||
AF486187.1 | - | 2 | 232 | |||
FQ221012.1 | - | 2 | 242 |