Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181384.2 | NP_852049.1 | 3 | 850 | ||
GenBank mRNA | AY259541.1 | - | 3 | 850 |