Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134858.3 | NP_001128330.1 | 4 | 4965 | ||
XM_006229337.2 | XP_006229399.1 | 5 | 4054 | |||
GenBank mRNA | AB091769.1 | - | 5 | 3967 |