Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SARDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134707.1 | NP_001128179.1 | 4, 5 | 824 | ||
NM_007101.3 | NP_009032.2 | 4, 5 | 983 | |||
XM_017014367.1 | XP_016869856.1 | 4, 5 | 986 | |||
XM_017014368.1 | XP_016869857.1 | 4, 5 | 986 | |||
GenBank mRNA | AF047004.1 | - | 4, 5 | 822 | ||
AF047190.1 | - | 4, 5 | 822 | |||
AF095735.2 | - | 4, 5 | 948 | |||
AF095736.2 | - | 3, 4 | 560 | |||
AF095737.2 | - | 4, 5 | 952 | |||
AF162428.1 | - | 3, 4 | 720 | |||
AJ223317.1 | - | 4, 5 | 822 | |||
AK223402.1 | - | 4, 5 | 870 | |||
AK291211.1 | - | 4, 5 | 818 | |||
AK298348.1 | - | 4, 5 | 867 | |||
AK316494.1 | - | 3, 4 | 622 | |||
BC136363.1 | - | 4, 5 | 801 | 9020350 | ||
BC136364.1 | - | 4, 5 | 801 | 9020351 | ||
BC144035.1 | - | 4, 5 | 819 | 9052555 |