Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smagp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182817.1 | NP_877969.1 | 4 | 255 | ||
XM_006242323.2 | XP_006242385.1 | 4 | 384 | |||
GenBank mRNA | AJ577837.1 | - | 4 | 255 | ||
BC101910.1 | - | 4 | 570 | 7933976 |