Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166102.1 | NP_001159574.1 | 3 | 312 | ||
NM_007103.3 | NP_009034.2 | 3 | 339 | |||
GenBank mRNA | AF053070.1 | - | 3 | 255 | ||
AF092131.1 | - | 3 | 265 | |||
AK055875.1 | - | 1 | 1010 | |||
AK223061.1 | - | 3 | 233 | |||
AK293522.1 | - | 2 | 150 | |||
AK300726.1 | - | 3 | 233 | |||
AK310537.1 | - | 3 | 243 | |||
AK316122.1 | - | 3 | 180 | |||
BC008146.1 | - | 3 | 224 | 4156629 | ||
BC015645.2 | - | 3 | 219 | 4648781 | ||
CR456739.1 | - | 3 | 186 |