Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182820.1 | NP_877972.1 | 4 | 295 | ||
XM_006247586.2 | XP_006247648.1 | 6 | 474 | |||
XM_006247588.2 | XP_006247650.1 | 6 | 473 | |||
XM_006247595.2 | XP_006247657.1 | 5 | 443 | |||
XM_008768359.1 | XP_008766581.1 | 6 | 474 | |||
XM_008768360.1 | XP_008766582.1 | 6 | 450 | |||
XM_008768361.1 | XP_008766583.1 | 4 | 386 | |||
XM_008768362.1 | XP_008766584.1 | 4 | 362 | |||
XM_008768363.1 | XP_008766585.1 | 5 | 444 | |||
XM_008768364.1 | XP_008766586.1 | 5 | 420 | |||
GenBank mRNA | AY327409.1 | - | 4 | 295 | ||
BC081911.1 | - | 5 | 379 | 7111907 |