Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhebl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182825.1 | NP_877977.1 | 3 | 321 | ||
XM_008765754.1 | XP_008763976.1 | 3 | 339 | |||
XM_008765755.1 | XP_008763977.1 | 3 | 339 | |||
GenBank mRNA | AY327411.1 | - | 3 | 321 |