Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183403.2 | NP_899653.2 | 2 | 422 | ||
GenBank mRNA | CX570255.1 | - | 2 | 121 |