Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnrh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012767.2 | NP_036899.1 | 4 | 379 | ||
XM_006252096.2 | XP_006252158.1 | 3 | 380 | |||
GenBank mRNA | M12579.1 | - | 4 | 379 |