Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286696.1 | NP_001273625.1 | 5 | 1934 | ||
NM_001286697.1 | NP_001273626.1 | 6 | 1936 | |||
NM_001286698.1 | NP_001273627.1 | 3 | 1711 | |||
NM_001322260.1 | NP_001309189.1 | 6 | 2065 | |||
NM_007135.2 | NP_009066.2 | 5 | 1827 | |||
XM_006717279.3 | XP_006717342.1 | 6 | 2035 | |||
GenBank mRNA | AK054606.1 | - | 5 | 1819 | ||
AK302284.1 | - | 5 | 1876 | |||
AK316233.1 | - | 6 | 1872 | |||
BC062309.1 | - | 5 | 1782 | 3944657 |