Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufaf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033971.1 | NP_001029143.1 | 3 | 986 | ||
NM_020080.2 | NP_064465.1 | 3 | 1004 | |||
XM_006243845.2 | XP_006243907.1 | 4 | 2486 | |||
GenBank mRNA | BC079306.1 | - | 3 | 738 | 7134644 | |
FQ216095.1 | - | 3 | 355 | |||
FQ223486.1 | - | 2 | 432 | |||
FQ233128.1 | - | 2 | 431 | |||
U95160.1 | - | 3 | 340 | |||
U95161.1 | - | 3 | 373 | |||
U95162.1 | - | 2 | 477 |