Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007553.2 | NP_001007554.1 | 7 | 1082 | ||
NM_001130523.2 | NP_001123995.1 | 7 | 1136 | |||
NM_001242891.1 | NP_001229820.1 | 8 | 1229 | |||
NM_001242892.1 | NP_001229821.1 | 6 | 695 | |||
NM_001242893.1 | NP_001229822.1 | 5 | 602 | |||
NM_007158.5 | NP_009089.4 | 6 | 989 | |||
GenBank mRNA | AB020692.1 | - | 7 | 996 | ||
AF070542.1 | - | 5 | 586 | |||
AF077054.1 | - | 6 | 896 | |||
AK022516.1 | - | 5 | 504 | |||
AK022528.1 | - | 5 | 504 | |||
AK290146.1 | - | 6 | 627 | |||
AY049788.1 | - | 4 | 491 | |||
BC032446.1 | - | 6 | 924 | 5111213 | ||
BX648561.1 | - | 6 | 826 | |||
BX648780.1 | - | 6 | 923 | |||
CR749378.1 | - | 7 | 1086 | |||
CR749431.1 | - | 7 | 1068 |