Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021593.1 | NP_067604.1 | 3 | 210 | ||
XM_006250325.2 | XP_006250387.1 | 3 | 375 | |||
GenBank mRNA | AF056031.1 | - | 3 | 210 | ||
BC088142.1 | - | 3 | 191 | 7322842 | ||
FQ210049.1 | - | 3 | 208 |