Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021594.1 | NP_067605.1 | 6 | 1230 | ||
GenBank mRNA | AF154336.1 | - | 6 | 1230 | ||
FQ213792.1 | - | 6 | 1347 | |||
FQ228926.1 | - | 6 | 1328 |