Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007167.3 | NP_009098.3 | 4 | 503 | ||
GenBank mRNA | AK092067.1 | - | 3 | 215 | ||
AK299293.1 | - | 4 | 462 | |||
AK301461.1 | - | 3 | 271 | |||
AK310703.1 | - | 4 | 444 | |||
AL389985.1 | - | 4 | 428 | |||
AL832911.1 | - | 4 | 449 | |||
BC033903.1 | - | 4 | 485 | |||
BC068534.1 | - | 4 | 503 | |||
BC107880.1 | - | 4 | 484 | |||
BC117463.1 | - | 4 | 371 | |||
BT007117.1 | - | 3 | 275 |