Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021853.1 | NP_068625.1 | 18 | 1749 | ||
XM_006233697.2 | XP_006233759.1 | 18 | 2057 | |||
XM_006233698.2 | XP_006233760.1 | 18 | 2057 | |||
XM_006233699.2 | XP_006233761.1 | 18 | 2057 | |||
XM_006233700.2 | XP_006233762.1 | 17 | 2015 | |||
XM_006233701.2 | XP_006233763.1 | 18 | 2057 | |||
XM_006233702.2 | XP_006233764.1 | 17 | 2015 | |||
XM_006233703.2 | XP_006233765.1 | 18 | 2057 | |||
XM_006233704.2 | XP_006233766.1 | 17 | 1937 | |||
XM_006233705.2 | XP_006233767.1 | 17 | 1934 | |||
XM_006233706.2 | XP_006233768.1 | 18 | 2057 | |||
XM_006233707.2 | XP_006233769.1 | 17 | 1932 | |||
XM_008761624.1 | XP_008759846.1 | 18 | 2047 | |||
GenBank mRNA | AF089730.1 | - | 18 | 1749 | ||
AY884213.1 | - | 17 | 1647 |