Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022193.1 | NP_071529.1 | 495 | |||
GenBank mRNA | EF121986.1 | - | 495 | |||
EF121987.1 | - | 495 | ||||
J03808.1 | - | 495 |