Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC60591 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022203.1 | NP_071539.1 | 3 | 334 | ||
GenBank mRNA | U79661.1 | - | 3 | 334 |