Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022214.1 | NP_071550.1 | 2 | 145 | ||
XM_008770053.1 | XP_008768275.1 | 9 | 1542 | |||
XM_008770054.1 | XP_008768276.1 | 2 | 562 | |||
XM_008770055.1 | XP_008768277.1 | 9 | 1539 | |||
GenBank mRNA | U90448.1 | - | 2 | 145 |