Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022215.2 | NP_071551.2 | 3 | 351 | ||
XM_006257376.1 | XP_006257438.1 | 4 | 553 | |||
GenBank mRNA | AB002558.1 | - | 3 | 339 | ||
BC088396.1 | - | 3 | 351 | 7108237 |