Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022234.2 | NP_071570.2 | 6, 7 | 1539 | ||
XM_006245275.2 | XP_006245337.1 | 7, 8 | 1668 | |||
GenBank mRNA | AJ242554.1 | - | 6, 7 | 1520 | ||
AJ271642.1 | - | 6, 7 | 1443 |