Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022277.1 | NP_071613.1 | 2, 3 | 616 | ||
XM_006245014.2 | XP_006245076.1 | 2, 3 | 598 | |||
XM_006245015.2 | XP_006245077.1 | 2, 3 | 739 | |||
XM_008767135.1 | XP_008765357.1 | 2, 3 | 598 | |||
GenBank mRNA | AF279308.1 | - | 2, 3 | 616 | ||
AF288372.1 | - | 2, 3 | 351 |