Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Safb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022394.1 | NP_071789.1 | 7 | 1229 | ||
GenBank mRNA | AF056324.1 | - | 7 | 1229 |
Gene Symbol: Safb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002730092.4 | 7 | 1303 | |||
XM_006244363.2 | XP_006244425.1 | 7 | 1303 | |||
XM_006244364.2 | XP_006244426.1 | 7 | 1303 | |||
XM_006244365.2 | XP_006244427.1 | 7 | 1303 | |||
XM_006244366.2 | XP_006244428.1 | 7 | 1303 | |||
XM_006244367.2 | XP_006244429.1 | 7 | 1303 | |||
XM_006244368.2 | XP_006244430.1 | 4 | 757 | |||
XM_008766801.1 | XP_008765023.1 | 7 | 1303 | |||
GenBank mRNA | FQ221088.1 | - | 7 | 1281 |