Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199214.1 | NP_001186143.1 | 4 | 786 | ||
NM_007029.3 | NP_008960.2 | 4 | 786 | |||
XM_005251142.2 | XP_005251199.1 | 5 | 410 | |||
GenBank mRNA | AK292737.1 | - | 4 | 458 | ||
AK297594.1 | - | 5 | 447 | |||
AK299500.1 | - | 4 | 375 | |||
AK308557.1 | - | 5 | 567 | |||
AK309130.1 | - | 4 | 673 | |||
AL110174.1 | - | 1 | 47 | |||
BC006302.2 | - | 4 | 455 | 4100671 | ||
BQ069488.1 | - | 4 | 448 | |||
BT020034.1 | - | 3 | 404 | |||
CR456833.1 | - | 3 | 404 | |||
D50375.1 | - | 4 | 521 | |||
S82024.1 | - | 4 | 432 |