Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022510.1 | NP_071955.1 | 6 | 678 | ||
GenBank mRNA | BC063811.1 | - | 6 | 694 | 6919340 | |
BC081801.1 | - | 6 | 649 | 7192390 | ||
FQ209448.1 | - | 5 | 532 | |||
FQ209532.1 | - | 5 | 532 | |||
FQ209571.1 | - | 5 | 532 | |||
FQ212557.1 | - | 5 | 494 | |||
FQ212566.1 | - | 5 | 532 | |||
FQ212656.1 | - | 5 | 532 | |||
FQ212694.1 | - | 650 | ||||
FQ212896.1 | - | 5 | 532 | |||
FQ212979.1 | - | 3 | 324 | |||
FQ213045.1 | - | 5 | 532 | |||
FQ213454.1 | - | 6 | 683 | |||
FQ215017.1 | - | 5 | 488 | |||
FQ215042.1 | - | 5 | 532 | |||
FQ215264.1 | - | 5 | 532 | |||
FQ215283.1 | - | 5 | 532 | |||
FQ215533.1 | - | 5 | 532 | |||
FQ215586.1 | - | 5 | 532 | |||
FQ215649.1 | - | 5 | 532 | |||
FQ216342.1 | - | 5 | 533 | |||
FQ216573.1 | - | 5 | 532 | |||
FQ216754.1 | - | 5 | 532 | |||
FQ216792.1 | - | 5 | 532 | |||
FQ218195.1 | - | 5 | 532 | |||
FQ219327.1 | - | 5 | 532 | |||
FQ219701.1 | - | 5 | 532 | |||
FQ219716.1 | - | 5 | 532 | |||
FQ220231.1 | - | 5 | 530 | |||
FQ220328.1 | - | 3 | 354 | |||
FQ221130.1 | - | 5 | 494 | |||
FQ221674.1 | - | 5 | 532 | |||
FQ226216.1 | - | 5 | 532 | |||
FQ228709.1 | - | 5 | 532 | |||
FQ228734.1 | - | 6 | 683 | |||
FQ228797.1 | - | 5 | 532 | |||
FQ228847.1 | - | 5 | 532 | |||
FQ228910.1 | - | 5 | 532 | |||
FQ228929.1 | - | 5 | 532 | |||
FQ228930.1 | - | 5 | 532 | |||
FQ229086.1 | - | 5 | 532 | |||
FQ229087.1 | - | 5 | 532 | |||
FQ229116.1 | - | 5 | 532 | |||
FQ229475.1 | - | 2 | 165 | |||
FQ229723.1 | - | 3 | 272 | |||
FQ229849.1 | - | 5 | 491 | |||
FQ231653.1 | - | 5 | 532 | |||
FQ232105.1 | - | 5 | 532 | |||
FQ232771.1 | - | 5 | 532 | |||
FQ233898.1 | - | 5 | 532 | |||
FQ234293.1 | - | 5 | 532 | |||
FQ234619.1 | - | 5 | 532 | |||
X82180.1 | - | 6 | 678 |