Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022546.1 | NP_071991.1 | 4 | 595 | ||
XM_006240991.1 | XP_006241053.1 | 4 | 726 | |||
XM_006240993.2 | XP_006241055.1 | 4 | 833 | |||
XM_008765129.1 | XP_008763351.1 | 4 | 1059 | |||
GenBank mRNA | AB010083.1 | - | 1 | 74 | ||
AJ006971.1 | - | 4 | 595 | |||
BC062076.1 | - | 4 | 596 | 5599435 |